Stampa…

N-Methylethylamine 97%

Fornitore: Thermo Scientific
Pericolo

Synonyms: N-Ethylmethylamine

314680050 314680250
ACRO314680050EA 71.3 EUR
ACRO314680050 ACRO314680250
N-Methylethylamine 97%
N-Methylethylamine
Formula: C₃H₉N
Peso molecolare: 59,11 g/mol
Punto di ebollizione: 33…34 °C (760 mmHg)
Densità: 0,68 g/cm³
Flash Pt: <–34 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00009030
CAS Number: 624-78-2
EINECS: 210-862-1
UN: 2733
ADR: 3,I

Visualizza i codici

Risultati del test di specificazione

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3730 to 1.3770 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR