Tetrabutylammonium tetrahydroborate 98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate
Formula:
C₁₆H₄₀BN Punto di fusione: 126…131 °C |
MDL Number:
MFCD00012035 CAS Number: 33725-74-5 EINECS: 251-658-2 UN: 3131 ADR: 4.3,II |
Risultati del test di specificazione
Appearance | White to off-white Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...