Stampa…

N-(Hydroxymethyl)phthalimide 97%

Fornitore: Thermo Scientific

Synonyms: 2-(Hydroxymethyl)isoindoline-1,3-dione

121490250 121491000
ACRO121490250EA 25.9 EUR
ACRO121490250 ACRO121491000
N-(Hydroxymethyl)phthalimide 97%
N-(Hydroxymethyl)phthalimide
Formula: C₉H₇NO₃
Peso molecolare: 177,16 g/mol
Punto di ebollizione: 415 °C (1013 hPa)
Punto di fusione: 140…145 °C
MDL Number: MFCD00005899
CAS Number: 118-29-6
EINECS: 204-241-4

Visualizza i codici

Risultati del test di specificazione

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 140 to 145 °C
Loss on drying ≤2 %
Sulfated ash ≤0.1 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR