Stampa…
(Benzyl)triphenylphosphonium chloride 99%
Fornitore: Thermo Scientific
Pericolo
Synonyms: Benzyltriphenylphosphonium chloride
106210250 106211000 106215000
ACRO106210250EA 31.9 EUR
ACRO106210250 ACRO106211000 ACRO106215000
(Benzyl)triphenylphosphonium chloride 99%
(Benzyl)triphenylphosphonium chloride
Formula: C₂₅H₂₂ClP
Peso molecolare: 388,88 g/mol
Punto di fusione: 337 °C
Densità: 1,18 g/cm³ (20 °C)
Flash Pt: >300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011913
CAS Number: 1100-88-5
EINECS: 214-154-3
UN: 3464
ADR: 6.1,II
Visualizza i codici
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
GC ≥94 %