Stampa…

Methyl hydrogen adipate 97%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 1,6-Hexanedioic acid methyl ester, Adipic acid methyl ester, Adipic acid monomethyl ester, 1,6-Hexanedioic acid monomethyl ester, Monomethyl adipate

102820050 102820250 102821000
ACRO102820050EA 17.7 EUR
ACRO102820050 ACRO102820250 ACRO102821000
Methyl hydrogen adipate 97%
Methyl hydrogen adipate
Formula: C₇H₁₂O₄
Peso molecolare: 160,17 g/mol
Punto di ebollizione: 162 °C (10 mmHg)
Punto di fusione: 8…9 °C
Densità: 1,08 g/cm³
Flash Pt: >109 °C
MDL Number: MFCD00004418
CAS Number: 627-91-8
EINECS: 211-019-0

Visualizza i codici

Risultati del test di specificazione

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20°C, 589 nm)
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR