2,5-Dimethylbenzenesulphonyl chloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₈H₉ClO₂S Punto di ebollizione: 150°C |
MDL Number:
MFCD00024875 CAS Number: 19040-62-1 UN: 3261 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...