(±)-Epicloridrina 99%
Fornitore: Apollo Scientific
Synonyms:
3-Cloro-1,2-epossipropano, (±)-1-Cloro-2,3-epossipropano, (±)-3-Cloro-1,2-epossipropano, 1-Cloro-2,3-epossipropano, Epicloridrina
OR922424-25G
OR922424-100G
OR922424-500G
APOSOR922424-25GEA
21.9
EUR
APOSOR922424-25G
APOSOR922424-100G
APOSOR922424-500G
(±)-Epicloridrina 99%
(±)-Epicloridrina
Formula:
C₃H₅ClO Peso molecolare: 92,52 g/mol |
MDL Number:
MFCD00005132 CAS Number: 106-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...