Stampa…
Methyl 3-hydroxy-5-nitrobenzoate 98%
Fornitore: Apollo Scientific
Attenzione
OR52897-1G OR52897-5G OR52897-10G
APOSOR52897-1GEA 125 EUR
APOSOR52897-1G APOSOR52897-5G APOSOR52897-10G
Methyl 3-hydroxy-5-nitrobenzoate 98%
Methyl 3-hydroxy-5-nitrobenzoate
Formula: C₈H₇NO₅
MDL Number: MFCD00463877
CAS Number: 55076-32-9