Stampa…
Methyl 2-chloro-6-fluoro-3-methoxybenzoate 95%
Fornitore: Apollo Scientific
Attenzione
PC57473-500MG PC57473-1G PC57473-5G PC57473-25G
APOSPC57473-500MGEA 251 EUR
APOSPC57473-500MG APOSPC57473-1G APOSPC57473-5G APOSPC57473-25G
Methyl 2-chloro-6-fluoro-3-methoxybenzoate 95%
Methyl 2-chloro-6-fluoro-3-methoxybenzoate
Formula: C₉H₈ClFO₃
Peso molecolare: 218,609 g/mol
MDL Number: MFCD10566180
CAS Number: 1379356-16-7