6-FAM phosphoramidite [5'-Fluorescein phosphoramidite] for oligonucleotide labeling
Fornitore: AAT BIOQUEST INC
|
Attenzione
|
Synonyms:
6-Fluorescein Phosphoramidite, 6-((6-(((2-Cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)hexyl)carbamoyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl bis(2,2-dimethylpropanoate), 5'-Fluorescein CE-Phosphoramidite (6-FAM)
There are several ways of labeling an oligonucleotide with fluorescein.
- FRET building block
- Multiple applications
Formula:
C₄₆H₅₈N₃O₁₀P Peso molecolare: 843,94 g/mol |
MDL Number:
MFCD01940925 CAS Number: 204697-37-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...