Stampa…
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥95%
Fornitore: AAT BIOQUEST INC
Attenzione
Synonyms: Bisbenzimide H 33258, 4-(5-(4-Methylpiperazin-1-yl)-1H,1'H-[2,5'-bibenzo[d]imidazol]-2'-yl)phenol trihydrochloride, 2'-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bibenzimidazole trihydrochloride hydrate
17520 17523
AATB17520EA 146 EUR
AATB17520 AATB17523
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥95%
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.
Formula: C₂₅H₂₄N₆O·3HCl·xH₂O
Peso molecolare: 533,88 g/mol (anhy)
MDL Number: MFCD00012679
CAS Number: 23491-45-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b6851d2f3cca5b4f1783f3c851b9c9fdc1727de&vid=ef3db57e4ef2a1c751ce3ada15e71ca058e37c8a&prid=web&clid=SAPDAM