Stampa…

Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 555)

Fornitore: AAT BIOQUEST INC
16460 16739
AATB16460EA 115 EUR
AATB16460 AATB16739
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 555)
Anticorpi
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Attenzione: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 555
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR