Stampa…
7-Methoxycoumarin-3-carboxylic acid ≥95%
Fornitore: AAT BIOQUEST
Attenzione
Synonyms: 7-Methoxy-2-oxo-2H-chromene-3-carboxylic acid
560 561
AATB560EA 230 EUR
AATB560 AATB561
7-Methoxycoumarin-3-carboxylic acid ≥95%
7-Methoxycoumarin-3-carboxylic acid
7-Methoxycoumarin-3-carboxylic acid is used for HPLC derivatization.
Formula: C₁₁H₈O₅
Peso molecolare: 220.18 g/mol
MDL Number: MFCD00452770
CAS Number: 20300-59-8
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=822555a76585018f1d64aa1962af306a89fbd630&vid=cd52cba0d62aab0731a70b771ec2c8f1fb6b2757&prid=web&clid=SAPDAM