Stampa…
Tide Quencher™ 2WS alkyne, Tide Quencher™ fluorescent probe
Fornitore: AAT BIOQUEST INC
Attenzione
Synonyms: TQ2WS alkyne
2213 2214
AATB2213EA 146 EUR
AATB2213 AATB2214
Tide Quencher™ 2WS alkyne, Tide Quencher™ fluorescent probe
Tide Quencher™ 2WS alkyne
TQ2WS is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.
  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling
Peso molecolare: 527,66 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d63636908f0a73fc395ab4df030d2c69ab26808&vid=8895c8742081d20d4a7e30554cdfdb476c7b3570&prid=web&clid=SAPDAM