Stampa…

ProLite™ Orange 5000X stain_protein gel

Fornitore: AAT BIOQUEST
Attenzione

18000 18001
AATB18000EA 137 EUR
AATB18000 AATB18001
ProLite™ Orange 5000X stain_protein gel
ProLite™ Orange

Prolite™ Orange is an alternative protein stain that can be used to replace SYPRO Orange Protein Gel Stain (SYPRO is the trademark of ThermoFisher).


  • Superior sensitivity
  • Compataible with all major viewing platforms
  • Rapid staining
  • Can be used for total protein quantitation
  • Alternative to sypro orange


Attenzione: For research use only.

Peso molecolare: 486.72 g/mol
ADR: 5 II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR