Stampa…
Eu-Cryptate succinimidyl ester, trFluor™ fluorescent probe
Fornitore: AAT Bioquest
Attenzione
1430 1431
AATB1430EA 460 EUR
AATB1430 AATB1431
Eu-Cryptate succinimidyl ester, trFluor™ fluorescent probe
Eu-Cryptate succinimidyl ester
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • No crosslinking
  • High stability and selectivity
  • Insensitive to fluorescence quenching
  • High emission yield
ADR: 5,II