Stampa…
FITC (fluoresceina 5-isotiocianato, fluoresceina isotiocianato isomero I) ≥90%
Fornitore: AAT BIOQUEST
Attenzione
Synonyms: 5-isothiocyanatofluorescein, Fluorescein 5(6)-isothiocyanate, Fluorescein Isothiocyanate, Isomer I
120 121 122
AATB120EA 152 EUR
AATB120 AATB121 AATB122
FITC (fluoresceina 5-isotiocianato, fluoresceina isotiocianato isomero I) ≥90%
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
5-FITC and 6-FITC have very similar absorption and fluorescence spectra.
Formula: C₂₁H₁₁NO₅S
Peso molecolare: 389.381 g/mol
MDL Number: MFCD00005063
CAS Number: 3326-32-7
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=866efe5f0d0dbbf2eb5a527d99d2c4423364c6ed&vid=a30e56d7f8123f0d56efdd7fe3ae64e2d98985c4&prid=web&clid=SAPDAM