Stampa…
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥90%
Fornitore: AAT BIOQUEST
Attenzione
Synonyms: 2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate, 5-FAM-N-succinimidyl ester, 5-Carboxyfluorescein NHS ester, 5-FAM SE
113 114 115
AATB113EA 152 EUR
AATB113 AATB114 AATB115
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥90%
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)
5-FAM, SE is the amine-reactive succinimidyl ester of single isomer 5-FAM acid.
Formula: C₂₅H₁₅NO₉
Peso molecolare: 473.39 g/mol
MDL Number: MFCD00077322
CAS Number: 92557-80-7
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47610eeaecfe0164a88fe381d6d7a09565d50115&vid=f3c00ee395340a181740bfb53be02a7ee9e47c81&prid=web&clid=SAPDAM