Stampa…

Polytetrahydrofuran

Fornitore: TCI

Synonyms: PTHF

P3027-25G P3027-500G
TCIAP3027-25GEA 29 EUR
TCIAP3027-25G TCIAP3027-500G
Polytetrahydrofuran
Polytetrahydrofuran

Store under inert gas.

Formula: H(C₄H₈O)nOH
MDL Number: MFCD00148879
CAS Number: 25190-06-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR