Zinc picolinate, in polvere 200-400 Mesh
Fornitore: Spectrum Chemical
Z1088-12KGBL
Z1088-1KG
Z1088-25KGBL
Z1088-50KGBL
SPCMZ1088-12KGBLEA
4600
EUR
SPCMZ1088-12KGBL
SPCMZ1088-1KG
SPCMZ1088-25KGBL
SPCMZ1088-50KGBL
Zinc picolinate, in polvere 200-400 Mesh
Zinc picolinate
Zinc Picolinate, 200 to 400 Mesh, Powder is most commonly used as a nutritional food supplement as a source for zinc and aspartic acid.
Temperatura di stoccaggio:
Ambiente |
MDL Number:
MFCD00145544 CAS Number: 17949-65-4 |
Risultati del test di specificazione
Assay | Aprx.200 MG Zn/G |
Lead (Pb) | max. 10 ppm |
Arsenic (As) | max. 5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...