Stampa…
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99% USP
Fornitore: Spectrum Chemical
Attenzione
Synonyms: THAM
TR149-125GM TR149-1KG
SPCMTR149-125GMEA 102 EUR
SPCMTR149-125GM SPCMTR149-1KG
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99% USP
Tris-(idrossimetil) amminometano (TRIS)
Tromethamine, USP is used as an emulsifying agent excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula: NH₂C(CH₂OH)₃
Peso molecolare: 121.14 g/mol
Punto di ebollizione: 357 °C (1013 hPa)
Punto di fusione: 169…172 °C
Densità: 1,328 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM
Visualizza i codici
Assay 99% Min.