Stampa…
Cesium nitrate, ultrapuro
Fornitore: Spectrum Chemical
Synonyms: Nitric acid cesium salt
C1489-10KGBL C1489-1KG
SPCMC1489-10KGBLEA 11460 EUR
SPCMC1489-10KGBL SPCMC1489-1KG
Cesium nitrate, ultrapuro
Cesio nitratoi
Cesium Nitrate, Ultrapure is used as a colorant, as an oxidizer, and in pyrotechnic compositions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula: CsNO₃
Peso molecolare: 194.91 g/mol
Punto di ebollizione: 737
Punto di fusione: 414
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00010963
CAS Number: 7789-18-6
UN: 1451
ADR: 5.1 III
Merck Index: 12,02057