Stampa…
Sodium thiosulphate pentahydrate ≥99%, cristalli BP, Ph. Eur., JP, USP
Fornitore: Spectrum Chemical
SO210-500GM SO210-50KGBL SO210-12KG SO210-2.5KG
SPCMSO210-500GMEA 130 EUR
SPCMSO210-500GM SPCMSO210-50KGBL SPCMSO210-12KG SPCMSO210-2.5KG
Sodium thiosulphate pentahydrate ≥99%, cristalli BP, Ph. Eur., JP, USP
Sodio tiosolfato pentaidrato
Sodium Thiosulfate, Pentahydrate, Crystal, USP, EP, BP, JP is used as a bleach neutraliser.
All Spectrum Chemical USP, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula: Na₂S₂O₃.5H₂O
Peso molecolare: 248.19 g/mol
Punto di fusione: 48 °C
Densità: 1,67 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844
Visualizza i codici
Assay 99% Min.