Stampa…
3-Bromo-4-(trifluoromethyl)phenacyl bromide ≥95%
Fornitore: Apollo Scientific
PC500781-250MG PC500781-1G PC500781-5G
APOSPC500781-250MGEA 257 EUR
APOSPC500781-250MG APOSPC500781-1G APOSPC500781-5G
3-Bromo-4-(trifluoromethyl)phenacyl bromide ≥95%
3-Bromo-4-(trifluoromethyl)phenacyl bromide
Formula: C₉H₅Br₂F₃O
Peso molecolare: 345,9386 g/mol
MDL Number: MFCD22491199
CAS Number: 1383625-02-2
UN: 3261