Stampa…
Methyl 3,5-difluoro-4-methoxy-benzoate ≥95%
Fornitore: Apollo Scientific
Synonyms: Methyl 3,5-difluoro-4-methoxybenzoate
PC53604-250MG PC53604-1G PC53604-5G
APOSPC53604-250MGEA 116 EUR
APOSPC53604-250MG APOSPC53604-1G APOSPC53604-5G
Methyl 3,5-difluoro-4-methoxy-benzoate ≥95%
Methyl 3,5-difluoro-4-methoxy-benzoate
Formula: C₉H₈F₂O₃
Peso molecolare: 202.15482 g/mol
MDL Number: MFCD11520390
CAS Number: 329-45-3