Stampa…
4-Bromo-2,5-bis(trifluoromethyl)benzyl alcohol ≥95%
Fornitore: Apollo Scientific
PC56684-250MG PC56684-1G
APOSPC56684-250MGEA 234 EUR
APOSPC56684-250MG APOSPC56684-1G
4-Bromo-2,5-bis(trifluoromethyl)benzyl alcohol ≥95%
4-Bromo-2,5-bis(trifluoromethyl)benzyl alcohol
Formula: C₉H₅BrF₆O
Peso molecolare: 323,02981 g/mol
MDL Number: MFCD15526890
CAS Number: 1260815-51-7