Stampa…
2-Bromo-3,5-dichloro-4-fluoroaniline ≥98%
Fornitore: Apollo Scientific
PC52718-250MG PC52718-1G PC52718-5G
APOSPC52718-250MGEA 165 EUR
APOSPC52718-250MG APOSPC52718-1G APOSPC52718-5G
2-Bromo-3,5-dichloro-4-fluoroaniline ≥98%
2-Bromo-3,5-dichloro-4-fluoroaniline
Formula: C₆H₃BrCl₂FN
Peso molecolare: 258,90312 g/mol
MDL Number: MFCD11505662
CAS Number: 1092350-32-7