Stampa…
5-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid ≥95%
Fornitore: Apollo Scientific
PC503020-250MG PC503020-1G PC503020-5G
APOSPC503020-250MGEA 183 EUR
APOSPC503020-250MG APOSPC503020-1G APOSPC503020-5G
5-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid ≥95%
5-Chloro-2-fluoro-4-(trifluoromethyl)benzoic acid
Formula: C₈H₃ClF₄O₂
Peso molecolare: 242,5548 g/mol
MDL Number: MFCD28752592
CAS Number: 1807210-92-9