Stampa…
5-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine ≥95%
Fornitore: Apollo Scientific
PC49676-250MG PC49676-1G
APOSPC49676-250MGEA 183 EUR
APOSPC49676-250MG APOSPC49676-1G
5-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine ≥95%
5-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine
Formula: C₁₂H₁₄BBrF₃NO₂
Peso molecolare: 351,96 g/mol
MDL Number: MFCD26520142
CAS Number: 1256360-49-2