Stampa…
1-(3-Bromo-4-chloro-2-fluorophenyl)-2,2,2-trifluoroethan-1-one ≥95%
Fornitore: Apollo Scientific
Synonyms: 1-(3-Bromo-4-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone
PC49479-250MG PC49479-1G
APOSPC49479-250MGEA 219 EUR
APOSPC49479-250MG APOSPC49479-1G
1-(3-Bromo-4-chloro-2-fluorophenyl)-2,2,2-trifluoroethan-1-one ≥95%
1-(3-Bromo-4-chloro-2-fluorophenyl)-2,2,2-trifluoroethan-1-one