Stampa…
Internal thread cryogenic vials
Fornitore: Corning
430487 430488 430489 430490 430491 430492 430656 431386 431416 431417 431418 431419 431420 431421
734-1834EA 353.77 EUR
734-1834 734-1835 734-1836 734-1837 734-1838 734-1839 734-1853 734-4196 479-1187 479-1188 479-1189 479-1190 479-1191 479-1192
Internal thread cryogenic vials
Vial
These vials are manufactured from polypropylene to withstand temperatures down to –196 °C.
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal
  • 2 ml self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
Attenzione: Do not use cryogenic vials for storage inMore Product Information