Sodio L(+)-ascorbato 99,0, powder USP
Fornitore: Spectrum Chemical
Synonyms:
Acido L(+)-Ascorbico sale sodico
Sodium Ascorbate, Powder, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₆H₇NaO₆ Peso molecolare: 198.11 g/mol Punto di fusione: 219 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00082340 CAS Number: 134-03-2 |
|
Risultati del test di specificazione
Assay (Dried Basis) | 99.0 - 101.0% |
pH of a 10% Solution | 7.0 - 8.0 |
Specific Rotation [A]D | +103° to +108° |
Loss On Drying | max. 0.25% |
Heavy Metals (As Pb) | max. 0.002% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...