Stampa…
Ammonio solfato ≥99%, granular FCC
Fornitore: Spectrum Chemical
Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
AM185-2.5KG AM185-12KG
SPCMAM185-2.5KGEA 190 EUR
SPCMAM185-2.5KG SPCMAM185-12KG
Ammonio solfato ≥99%, granular FCC
Ammonio solfato
Ammonium Sulfate, Granular, FCC is used as an acidity regulator in flours and breads. It is listed on the FDA's Generally Recognised As Safe (GRAS) list of substances. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula: (NH₄)₂SO₄
Peso molecolare: 132.14 g/mol
Punto di fusione: 336…339 °C
Densità: 1.78 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559
Visualizza i codici
Assay 99% Min.