Stampa…
Respirators, gas filters, PX5 PAPR Series
Fornitore: RPB SAFETY
03-802-FR-EUUK 03-893-A2 03-895-ABEK
RPBS03-802-FR-EUUKEA 0 EUR
RPBS03-802-FR-EUUK RPBS03-893-A2 RPBS03-895-ABEK
Respirators, gas filters, PX5 PAPR Series
Respiratori Respiratori
The RPB PX5 is a battery powered air purifying respirator fan unit which draws in contaminated air from the wearer's immediate vicinity, passing it through internal filters, and supplying the filtered air to an approved RPB loose fitting respirator head top.
  • Backpack option available for applications that require a greater range of movement and work across varied angles
  • With a completely sealed housing, the PX5 has been certified to IP 65 rating in use and certified to IP 67 rating with cleaning kit in place
  • Filter door cannot mount to the unit unless a filter is fixed in place
  • HEPA filter removes up to 99,97% of particulates down to 0,12 µm
  • Gas filter protects against organic vapor exposure (replace pre-filter daily to extend the life of main filter)
  • Visual, audible, and vibrating alarms for low battery level, or when airflow falls below the minimum requirements
  • Long-life lithium-ion battery is protected from moisture when decontaminating and will not disconnect or bump off while operating
  • Operating temperature: −20 to +50 °C
  • Noise rating: speed 1: 57 dBA, speed 2: 59 dBA, speed 3: 62 dBA
  • Flow rate: speed 1: 170 L/min, speed 2: 210 L/min, speed 3: 240 L/min
Weighing just 1,16 kg with HEPA filter fitted (1,65 to 1,7 kg with gas filter fitted), the sleek, lightweight, compact design maximizes comfort. PX5 unit attaches to a flexi-belt system (fitted with the decon easy clean belt) via an intuitive vertical mount lock system (easily removes for quick and simple decontamination).
Deigned to be as compact and comfortable as possible, the Easy clean PX5 Starter kit is perfect for use within infection control, and cleanroom environments, where easily and effectively cleaning the system is of utmost importance. Depending on what type of contaminant users require protection from, the PX5 has various filter options, including gas and multi filters. The PX5 also has built-in audible, visual, and physical alarms to warn the wearer when their filter needs to be changed, or when there is only 5% of the battery charge remaining.
The unit is IP 65 (in use) and IP 67 rated.
Certificazioni: EN 12941:1998+A2:2008 TH3/2 P R SL.
Informazioni per l' accessorio: RPB PX5 PAPR Breathing Tube for Z-Link, T-Link, T200, Z4, and Nova 3 respirator ranges. RPB PX5 PAPR Short Breathing Tube for Z-Link, T-Link, T200, Z4, and Nova 3 respirator ranges when used with RPBs range of PAPR back harness (03-822-DC and 03-822-FR).
Informazioni per l'ordine: Includes 1×PX5 PAPR, 1×FR belt, 1×gas Door, 1×pack of 10 prefilters, 1×lithium ion rechargeable battery, 1×charging bank with EUUK plugs and 1×
flow tester. Note: head top, gas filter and breathing tube sold separately.