Stampa…
Cleanroom overalls, Tyvek® IsoClean®, Model IC193B TS
Fornitore: DuPont
IC193BWHTS/XS IC193BWHTS/S IC193BWHTS/M IC193BWHTS/L IC193BWHTS/XL IC193BWHTS/2XL IC193BWHTS/3XL IC193BWHTS/4XL IC193BWHTS/5XL IC193BWHTS/6XL IC193BWHTS/7XL
DUPOIC193BWHTS/XSEA 535 EUR
DUPOIC193BWHTS/XS DUPOIC193BWHTS/S DUPOIC193BWHTS/M DUPOIC193BWHTS/L DUPOIC193BWHTS/XL DUPOIC193BWHTS/2XL DUPOIC193BWHTS/3XL DUPOIC193BWHTS/4XL DUPOIC193BWHTS/5XL DUPOIC193BWHTS/6XL DUPOIC193BWHTS/7XL
Cleanroom overalls, Tyvek® IsoClean®, Model IC193B TS
Indumenti usa e getta
These hooded coveralls, model IC193BWH TS, are made of Tyvek®, a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Hooded overall
  • Integrated overboots with a slip retardant Gripper™ sole
  • Ultra-low linting fabric
  • Tyvek® covered elasticated thumb loops
  • Tunnelled elastic at wrists and ankles; stitched-in elastic waist
  • Front zipper closure with storm flap
  • Optimised shape and fit allowing full freedom of movement
  • Aseptically folded
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments made of Tyvek® IsoClean® are used in the biotech, pharmaceutical, medical device manufacturing, and electronics industries, as well as in other critical or controlled environments.
Certificazioni: Type 5B/6B, EN 1073-2 (Class 2), EN 14126, Cat. III.
Confezione: Individually double packed. Two polyethylene liners. Cardboard box. Packed in an ISO Class 5 certified cleanroom.