Stampa…
Autoclave Bags with Steam Indicator, Self-Seal, DuPont™ Tyvek® 1422A
Fornitore: KEYSTONE CLEANROOM PRODUCTS
BGT2A1234SS BGT2A0416SS BGT2A0409SS BGT2A0205SS BGT2A0404SS BGT2A0426SS BGT2A0428SS BGT2A0430SS BGT2A0608SS BGT2A0610SS BGT2A0612SS BGT2A0615SS BGT2A0616SS BGT2A0638SS BGT2A0707SS BGT2A0709SS BGT2A0713SS BGT2A0811SS BGT2A0815SS BGT2A0820SS BGT2A09-512SS BGT2A1015SS BGT2A1018SS BGT2A1020SS BGT2A1022SS BGT2A1024SS BGT2A1030SS BGT2A1048SS BGT2A1056SS BGT2A1062SS BGT2A1212SS BGT2A1214SS BGT2A1216SS BGT2A1218SS BGT2A1220SS BGT2A1222SS BGT2A1224SS BGT2A1228SS BGT2A1230SS BGT2A1232SS BGT2A1236SS BGT2A1240SS BGT2A1242SS BGT2A1246SS BGT2A1248SS BGT2A1414SS BGT2A1416SS BGT2A1418SS BGT2A1420SS BGT2A1424SS BGT2A1426SS BGT2A1436SS BGT2A1438SS BGT2A1440SS BGT2A1448SS BGT2A1526SS BGT2A1543SS BGT2A1604SS BGT2A1612SS BGT2A1616SS BGT2A1620SS BGT2A1624SS BGT2A1630SS BGT2A1632SS BGT2A1636SS BGT2A1638SS BGT2A1644SS BGT2A1648SS BGT2A1820SS BGT2A1824SS BGT2A1830SS BGT2A1832SS BGT2A1836SS BGT2A1840SS BGT2A1848SS BGT2A1854SS BGT2A1870SS BGT2A2020SS BGT2A2024SS BGT2A2030SS BGT2A2032SS BGT2A2036SS BGT2A2040SS BGT2A2044SS BGT2A2046SS BGT2A2048SS BGT2A2224SS BGT2A2424SS BGT2A2430SS BGT2A2436SS BGT2A2440SS BGT2A2448SS BGT2A2452SS BGT2A2462SS BGT2A2628SS BGT2A2634SS BGT2A2646SS BGT2A2828SS BGT2A2832SS BGT2A2834SS BGT2A2844SS BGT2A3016SS BGT2A3020SS BGT2A3028SS BGT2A3030SS BGT2A3036SS BGT2A3060SS BGT2A3232SS BGT2A3248SS BGT2A3412SS BGT2A3434SS BGT2A3456SS BGT2A3624SS BGT2A3628SS BGT2A3636SS BGT2A3642SS BGT2A3646SS BGT2A3648SS BGT2A3660SS BGT2A3664SS BGT2A3672SS BGT2A3838SS BGT2A4024SS BGT2A4040SS BGT2A4048SS BGT2A4060SS BGT2A4072SS BGT2A4242SS BGT2A4244SS BGT2A4416SS BGT2A4450SS BGT2A4848SS BGT2A4850SS BGT2A4864SS BGT2A5054SS BGT2A6250SS BGT2A3427SS
129-3620EA 2640 EUR
129-3620 129-3162 129-3166 129-3194 129-3169 129-3155 129-3159 129-3161 129-3258 129-3262 129-3264 129-3266 129-3268 129-3271 129-3274 129-3276 129-3280 129-3284 129-3288 129-3291 129-3229 129-3236 129-3238 129-3241 129-3242 129-3245 129-3248 129-3250 129-3251 129-3253 129-3397 129-3592 129-3595 129-3598 129-3602 129-3608 129-3611 129-3613 129-3615 129-3618 129-3623 129-3626 129-3628 129-3546 129-3549 129-3556 129-3558 129-3562 129-3569 129-3574 129-3577 129-3579 129-3581 129-3583 129-3585 129-3588 129-3666 129-3668 129-3670 129-3672 129-3675 129-3678 129-3680 129-3692 129-3694 129-3695 129-3689 129-3687 129-3684 129-3631 129-3634 129-3636 129-3639 129-3641 129-3645 129-3647 129-3650 129-3653 129-3656 129-3661 129-3663 129-3433 129-3436 129-3439 129-3441 129-3443 129-3446 129-3452 129-3454 129-3457 129-3460 129-3464 129-3465 129-3400 129-3403 129-3405 129-3407 129-3411 129-3413 129-3415 129-3419 129-3422 129-3424 129-3426 129-3429 129-3431 129-3502 129-3505 129-3508 129-3510 129-3515 129-3517 129-3519 129-3523 129-3526 129-3533 129-3535 129-3540 129-3544 129-3466 129-3468 129-3470 129-3471 129-3474 129-3475 129-3477 129-3478 129-3480 129-3482 129-3484 129-3486 129-3491 129-3493 129-3497 129-2798 129-2800 KACCBGT2A3427SS
Autoclave Bags with Steam Indicator, Self-Seal, DuPont™ Tyvek® 1422A
Sacchetti Sacche di sterilizzazione
Self-sealing Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilisation of pharmaceutical change parts.
  • Manufactured and packed in a controlled cleanroom environment
  • Convenient self-sealing closure system
  • Tear and puncture resistant, hydrophobic and lint-free
  • Superior to paper-based autoclave pouches and wraps
  • Fast and consistent for equipment preparation area
  • Strong factory seals
  • Exceptional microbial barrier
Standardise and simplify the way equipment is wrapped for steam sterilisation. The use of these self-sealing bags will also save valuable time in the equipment prep area.
The drape able nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where you may need to 'feel through' the bag; especially when lifting or handling the wrapped part.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficiency and throughput.
Certificazioni: Certificates of Conformance sent with each shipment.