Stampa…
Masterflex® Double-Open-End Sediment Filter Cartridges
Fornitore: Avantor Fluid Handling
01512-00 01512-08 01512-18 01512-32 01512-38
MFLX01512-00EA 21.9 EUR
MFLX01512-00 MFLX01512-08 MFLX01512-18 MFLX01512-32 MFLX01512-38
Masterflex® Double-Open-End Sediment Filter Cartridges
Filtri Cartucce con filtro
Polypropylene fiber medium is thermally bonded to a rigid core for added durability.
  • Constructed of polypropylene (PP)
Ideal for removing a broad range of deformable and nondeformable particles. Rigid design is ideal for filtering high-solids, higher-viscosity fluids that require higher differential pressures. Cartridge imparts no taste, odor, or color. Maximum temperature is 79 °C (175 °F).
Note: For optimal performance, change cartridges at 25 psid.