Stampa…
Multiple distributors for bottles with GL 45 thread, BOLA
Fornitore: Bohlender
D614-08 D615-08
201-1710EA 97.2 EUR
201-1710 201-0299
Multiple distributors for bottles with GL 45 thread, BOLA
Ghiere Tappi per bottiglie
Tappo a vite in PPS, nero, corpo distributore in PTFE o PFA
  • Ottima resistenza chimica
  • Resistenza termica da −200 a +250 °C
  • Adatti per applicazioni con vuoto
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with a maximum external diameter of  8,5 mm can be inserted through the necks.