Stampa…
Laboratory refrigerators
Fornitore: Liebherr
LFKU1610 LFKU1612 LFKV3910 LFKV3912 LFKV5410B LFKU1613 LFKV3913 LFKU1653
471-0645EA 0 EUR
471-0645 471-0646 471-0648 471-0649 471-0653 471-1121 471-1122 LIEBLFKU1653
Laboratory refrigerators
Refrigeratori
Laboratory refrigerators with ventilated cooling at +5 °C ±3 °C, Cofrac certified.
  • Refrigeration system with automatic defrosting and condensation evaporation
  • Digital temperature regulation and display
  • Safety anti-freeze thermostat, audible door and temperature alarm
  • White epoxy-treated stainless steel body
  • Side compartment passage with a diameter of 30 mm
  • Housing for PT100 sensor, for centralised temperature control (without sensor)
  • RS 485 connection for control and traceability on a PC
Temperature range: +2 to +12 °C
Housing for PT100 sensor, for centralised temperature control (without sensor)
RS 485 connection for control and traceability on a PC