Desmopressin Acetate ≥99%
Fornitore: Apollo Scientific
BISN0004-5MG
BISN0004-25MG
BISN0004-50MG
APOSBISN0004-5MGEA
161
EUR
APOSBISN0004-5MG
APOSBISN0004-25MG
APOSBISN0004-50MG
Desmopressin Acetate ≥99%
Desmopressin Acetate
Desmopressin acetate is the synthetic analogue of the antidiuretic hormone arginine vasopressin.
Formula:
C₄₆H₆₄N₁₄O₁₂S₂.₂CH₃CO₂H Peso molecolare: 1189,321 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00133962 CAS Number: 62288-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...