Stampa…

Arabinofuranosyl-adenine ≥99%

Fornitore: Apollo Scientific

BIMN2036-1G BIMN2036-10G
APOSBIMN2036-1GEA 155 EUR
APOSBIMN2036-1G APOSBIMN2036-10G
Arabinofuranosyl-adenine ≥99%
Arabinofuranosyl-adenine
Formula: C₁₀H₁₃N₅O₄
Peso molecolare: 267,24 g/mol
Temperatura di stoccaggio: Ambiente
CAS Number: 2006-02-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR