Stampa…
D-Glucofuranuronic acid, gamma-lactone-1,6-13C2 Isotope_(99% 13C)
Fornitore: Apollo Scientific
BICL4106-100MG BICL4106-250MG BICL4106-500MG
APOSBICL4106-100MGEA 393 EUR
APOSBICL4106-100MG APOSBICL4106-250MG APOSBICL4106-500MG
D-Glucofuranuronic acid, gamma-lactone-1,6-13C2 Isotope_(99% 13C)
D-Glucofuranuronic acid, gamma-lactone-1,6-13C2