Stampa…
Photometric tube tests, NANOCOLOR® cyanide
Fornitore: MACHEREY-NAGEL
985031 985025
MANA985031EA 71.2 EUR
MANA985031 MANA985025
Photometric tube tests, NANOCOLOR® cyanide
Kit per analisi ambientali
A pre-dosed NANOCOLOR tube test for photometric determination of cyanide in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.