Stampa…

2',5'-Difluoroacetanilide

Fornitore: Apollo Scientific

Synonyms: N-(2,5-Difluorophenyl)acetamide

PC9973-1G PC9973-5G
APOSPC9973-1GEA 55.3 EUR
APOSPC9973-1G APOSPC9973-5G
2',5'-Difluoroacetanilide
2',5'-Difluoroacetanilide
Formula: C₈H₇F₂NO
Peso molecolare: 171,1441 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00032512
CAS Number: 398-90-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR