α,3-Dibromo-5-fluorotoluene 98%
Fornitore: Apollo Scientific
|
Pericolo
|
Formula:
C₇H₅Br₂F Flash Pt: 106 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD09258959 CAS Number: 216755-57-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...