Stampa…

3-Bromo-2-fluoro-5-methylpyridine 97%

Fornitore: Apollo Scientific

Synonyms: 3-Bromo-2-fluoro-5-picoline

PC9195-100G PC9195-10G PC9195-25G
APOSPC9195-100GEA 412 EUR
APOSPC9195-100G APOSPC9195-10G APOSPC9195-25G
3-Bromo-2-fluoro-5-methylpyridine 97%
3-Bromo-2-fluoro-5-methylpyridine
Formula: C₆H₅BrFN
Peso molecolare: 190,01 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03095305
CAS Number: 17282-01-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR