Stampa…

Fmoc-3-(trifluoromethyl)-L-phenylalanine 95+%

Fornitore: Apollo Scientific

Synonyms: Fmoc-Phe(3-CF₃)-OH, Fmoc-Phe(3-CF3)-OH

PC9136-1G PC9136-5G
APOSPC9136-1GEA 91.6 EUR
APOSPC9136-1G APOSPC9136-5G
Fmoc-3-(trifluoromethyl)-L-phenylalanine 95+%
Fmoc-3-(trifluoromethyl)-L-phenylalanine
Formula: C₂₅H₂₀F₃NO₄
Peso molecolare: 455,44 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00672555
CAS Number: 205526-27-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR