Stampa…

N-Ethyl-4-fluorobenzylamine 95%

Fornitore: Apollo Scientific

PC908738-1G PC908738-25G PC908738-5G
APOSPC908738-1GEA 125 EUR
APOSPC908738-1G APOSPC908738-25G APOSPC908738-5G
N-Ethyl-4-fluorobenzylamine 95%
N-Ethyl-4-fluorobenzylamine
Formula: C₉H₁₂FN
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04537942
CAS Number: 162401-03-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR