2-Fluoro-6-methylbenzoyl chloride 95%
Fornitore: Apollo Scientific
PC905471-1G
PC905471-5G
APOSPC905471-1GEA
132
EUR
APOSPC905471-1G
APOSPC905471-5G
2-Fluoro-6-methylbenzoyl chloride 95%
2-Fluoro-6-methylbenzoyl chloride
Formula:
C₈H₆ClFO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD06660278 CAS Number: 535961-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...