Stampa…

(4-Fluorophenyl)(oxo)acetonitrile 95%

Fornitore: Apollo Scientific

Synonyms: 4-Fluorobenzoyl cyanide

PC905326-1G PC905326-5G
APOSPC905326-1GEA 205 EUR
APOSPC905326-1G APOSPC905326-5G
(4-Fluorophenyl)(oxo)acetonitrile 95%
(4-Fluorophenyl)(oxo)acetonitrile
Formula: C₈H₄FNO
Peso molecolare: 149,13 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06658451
CAS Number: 658-13-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR